Orders LinkContactUsLinkComponent
Rudiger® Dentition Skull
Rudiger® Dentition Skull
Dentition Skull, 4, 18 cm, 25 cm, 18 cm
Catalog # 470015-744
Número de proveedor #:  A220
Rudiger® Dentition Skull
Catalog # 470015-744
Rudiger® Dentition Skull
Rudiger® Dentition Skull
Dentition Skull, 4, 18 cm, 25 cm, 18 cm
Price
$4,263.54
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
This skull features detailed teeth and a jaw.
  • Window Shows Dentition and Removable Teeth
  • Five-Year Manufacturer's Warranty
  • Individual Teeth Have Complete Roots
  • Removable Jaw
It's cast from a select natural bone specimen for unparalleled realism. Individual teeth have complete roots, and unlike the set of fused dentures found in other synthetic skulls, each tooth is in its own socket. A hinged flap in the right mandible swings open to expose the roots of the two permanently anchored premolars, first two molars in their sockets, and an impacted wisdom tooth.
A permanently flexible cement secures all the remaining teeth in the upper and lower jaws into their sockets, which allows users to extract teeth individually for thorough examination and then easily reinsert them. Once replaced in their sockets, teeth do not fall out, yet the unprecedented extraction procedure can be repeated numerous times. Detachable springs also permit natural articulation of the mandible. The jaw is removable, and the sectioned calvaria detaches for access to the neuro-cranium.
Ordering information: This product includes a manufacturer's warranty.
Especificaciones
  • Description:
    Dentition Skull
  • Height:
    18 cm
  • Length:
    25 cm
  • Width:
    18 cm
  • No. of Parts:
    4