Orders LinkContactUsLinkComponent
Color-Coded Inoculating Loops, Dynalon
Color-Coded Inoculating Loops, Dynalon
Color-Coded Inoculating Loops, Blue, w/0.5 mm dia. loop
  470147-476
 :   DYNALON
Supplier #:  226505
Color-Coded Inoculating Loops, Dynalon
  470147-476
 :  DYNALON
Color-Coded Inoculating Loops, Dynalon
Color-Coded Inoculating Loops, Dynalon
Color-Coded Inoculating Loops, Blue, w/0.5 mm dia. loop
$82.77
Paquete de 20 Case de 50 (20UN)
Made of color-coded styrene, these inoculating loops are easy to organize.
  • Two Sizes
  • Color-Coded Styrene to Distinguish Loop Size
  • Sturdy, 7½" Handles
  • Twenty Loops per Package
Both sizes of loops have sturdy, 7½" handles.
The handles and color-coding make these loops convenient and simple to use.
Packaging: One package has 20 loops.
  • Description:
    Color-Coded Inoculating Loops, Blue, w/0.5 mm dia. loop