Color Mixing Apparatus
Catalog # 470039-942
Supplier: UNITED SCIENTIFIC SUPPLIES
Specifications
- Descripción:Color Mixing Apparatus
- Referencia:470039-942
- Nº de proveedor:CMA001
Specifications
About this item
Illuminate the variances between additive and subtractive color mixing
- Independent control for each color
- Investigate additive and subtractive color mixing
A compact and economical Color Mixing Apparatus. This updated version of the classic three color light projector uses three low voltage, high power LED's to project easily-seen red, green, and blue light onto a white screen. The projector mounting allows the beam angles to be adjusted to vary the overlap of the three colored images. The intensity of each LED can be adjusted separately by a potentiometer to give a convincing white light or various shades of mixed colors. Colored shadows can also be explored in a darkened room. The set includes a wall mount power supply and activity.