Colon, Mammal, Thin Section Slide
Colon, Mammal, Thin Section, Thin section, Hematoxylin and Eosin
Catalog # 470183-688
Supplier: Avantor
Jump to:
Product Details & Documents
Extremely Thin Plastic Embedded Tissue For Greater Viewing Clarity
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Through the large intestine, showing absence of villi and changes in muscular layers. Goblet cells are numerous.
Through the large intestine, showing absence of villi and changes in muscular layers. Goblet cells are numerous.
Specifications
- Description:Colon, Mammal, Thin Section
- Stain:Hematoxylin and Eosin
- Orientation:Thin section