Colon Slide
Colon, Cross section, Hematoxylin and Eosin
Catalog # 470177-128
Supplier: Avantor
Jump to:
Product Details & Documents
Illustrates a Segment of the Digestive Tract
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Through the large intestine, showing absence of villi and changes in muscular layers. Goblet cells are numerous.
Specifications
- Description:Colon
- Stain:Hematoxylin and Eosin
- Orientation:Cross section