Orders LinkContactUsLinkComponent
Collision Track
Collision Track
Supplier:   VWR International
Collision Track
Supplier:  VWR International
Certificates
Collision Track
2 Options Available Below
Product Details & Documents
This track includes everything needed to demonstrate three basic concepts of physics: acceleration, collision, and force.
  • Instructions Provided
  • Two Carts Provided
  • Two Sets of Slotted Masses Provided
  • Two Photogate Support Rods Provided
  • Physics Topics Include Acceleration, Collision, and Force
Change the speed of the cart by using the steel slotted masses, or accelerate it by using the provided pulley to demonstrate the use of external forces.
The provided carts have spring bands on one side for elastic collisions and Velcro on the other for inelastic collisions.
Ordering information: This track includes a 1.2 m track, two carts, a pulley, two sets of slotted masses (10 g, 20 g, and 50 g), two photogate support rods, and instructions.
Product Family Options
Información del producto
  • Availability
    Price
  • 470017-142
    Collision Track
    Collision Track
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $2,211.98
    $2,211.98
    Each (1UN)
       $2,211.98
  • 470128-702
    Motorized Cart
    Motorized Cart
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $26.65
    $26.65
    Liquidación
    Each (1UN)
       $26.65