Collection Boxes
Acerca de este artículo
Create your own collections with these study boxes. Complete with styrene inserts and lids. Choose how many compartments you want.
- Three box sizes available
WL7075L-09
9 Cell Box
Box: 7 x 4.75 inches
Cells: 1.25 x 2 inches
WL7075L-15
15 Cell Box
Box: 11 x 7 inches
Cells: 1.75 x 1.75 inches
WL7075-24
24 Cell Box
Box: 11 x 7 inches
Cells: 1.5 x 1.25 inches
1 Options Available Below
Product Details & Documents
Create your own collections with these study boxes. Complete with styrene inserts and lids. Choose how many compartments you want.
- Three box sizes available
WL7075L-09
9 Cell Box
Box: 7 x 4.75 inches
Cells: 1.25 x 2 inches
WL7075L-15
15 Cell Box
Box: 11 x 7 inches
Cells: 1.75 x 1.75 inches
WL7075-24
24 Cell Box
Box: 11 x 7 inches
Cells: 1.5 x 1.25 inches
Product Family Options
Información del producto
- AvailabilityPrice
- 470151-142Collection Boxes, 9 cellSupplier Number: 0145-001
- 470151-140Collection Boxes, 15 cellCollection Boxes, 15 cellDiscontinued-Supplier Number: 0140-001
- 470151-138Collection Boxes, 24 cellCollection Boxes, 24 cellDiscontinued-Supplier Number: 0139-00