Orders LinkContactUsLinkComponent
Ward's® Coal (Anthracite)
 :   Avantor
Ward's® Coal (Anthracite)
 :  Avantor
Ward's® Coal (Anthracite)
Ward's® Coal (Anthracite)
Lustrous, black 'hard coal'.
  • Collected in Pennsylvania
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student Specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack - Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
  • Description
    Specimen Dimensions
  • Coal (Anthracite), student specimens
    1×1" - 1×1"
    Paquete de 10
    Paquete de 10 - $227.38
    $227.38
    Paquete de 10
       $227.38
  • Coal (Anthracite), hand specimen
    2×3" - 3×4"
    Each (1UN)
    Each (1UN) - $241.70
    $241.70
    Each (1UN)
       $241.70
  • Coal (Anthracite), bulk pack, 1 kg
    Size will vary
    Each (1UN)
    Each (1UN) - $324.18
    $324.18
    Each (1UN)
       $324.18