Orders LinkContactUsLinkComponent
<i>Clostridium rubrum</i> Slide
Clostridium rubrum Slide
Clostridium rubrum slide, Smear, Gram (+)
Catalog # 470180-894
Supplier:   Avantor
<i>Clostridium rubrum</i> Slide
Catalog # 470180-894
Supplier:  Avantor
<i>Clostridium rubrum</i> Slide
Clostridium rubrum Slide
Clostridium rubrum slide, Smear, Gram (+)
Price
$168.40
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Demonstrates Well-defined Endospore
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
An aerobic, spore forming, rod-shaped bacterium. Spores are oval in shape, and subterminal in location within the cell. This sometimes makes the cells appear to be ‘fatter’ at one end than the other.
Especificaciones
  • Description:
    Clostridium rubrum slide
  • Stain:
    Gram (+)
  • Orientation:
    Smear