Orders LinkContactUsLinkComponent
Closed Case Form Electroscope
Supplier:   VWR International
Closed Case Form Electroscope
Supplier:  VWR International
Certificates
Closed Case Form Electroscope
Closed Case Form Electroscope
2 Options Available Below
Product Details & Documents
Closed Case Form Electroscope is enclosed, preventing air currents from affecting the deflection of the leaves.
  • Booklet of Six Leaves
  • Instructions
  • Gold and Aluminum Leaf Construction
  • Enclosed Electroscope
  • Leaves Pivot when Charged
Leaves are a combination of gold and aluminum developed to provide strength without sacrificing sensitivity. Each leaf is suspended from two small hooks for simple installation, and the leaves pivot, rather than bend, when charged.
The metal case eliminates deflections caused by charges residing on glass walls. An insulating plastic sleeve holds the leaf-support rod in place. Front and rear sides of the box are made of a 10 cm square glass pane to allow visibility and shadow projection.
Ordering information: This kit includes a booklet of six leaves and instructions.
Product Family Options
Información del producto
  • Availability
    Price
  • 470148-720
    Closed Case Form Electroscope
    Closed Case Form Electroscope
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $553.57
    $553.57
    Log in Para disponibilidad
    Case de 40 (1UN)
    Case de 40 (1UN) - $603.39
    $603.39
    Each (1UN)
       $553.57
    Case de 40 (1UN)
       $603.39
  • 470149-480
    Replacement Foil for Closed Case Form Electroscope
    Replacement Foil for Closed Case Form Electroscope
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 6
    Paquete de 6 - $138.12
    $138.12
    Log in Para disponibilidad
    Case de 50 (6UN)
    Case de 50 (6UN) - $150.56
    $150.56
    Paquete de 6
       $138.12
    Case de 50 (6UN)
       $150.56