Clay Triangles
Clay Triangles, 2"
Catalog # 470176-154
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: CPT020
Jump to:
Product Details & Documents
Clay pipe triangle for holding items over a flame.
- Heat resistant clay pipes
Galvanized, 16-gauge iron wire with plain clay-pipe stems.
Specifications
- Description:Clay Triangles, 2"