Orders LinkContactUsLinkComponent
Large Magnetic Demonstration Prism Set
Large Magnetic Demonstration Prism Set
  470002-942
 :  PST620
Large Magnetic Demonstration Prism Set
  470002-942
Large Magnetic Demonstration Prism Set
Large Magnetic Demonstration Prism Set
Large Magnetic Demonstration Prism Set
$5,055.57
Each (1UN)
Teach a wide variety of optical principles with this set of seven lenses and prisms
  • Durable storage box
  • Magnetic backing on optical components
An oversized set of 6 lenses and prisms designed for mounting on a magnetic chalk or marker board. Lenses are about 8" long, so your optics experiments would be clearly visible from every part of the classroom. Each piece in the set has a pre-attached magnetic backing. An activity guide is also included. Set comes in a sturdy wooden storage case. Please note: Laser Ray Box not included.
 : Contents include Semi-circular lens; Equilateral prism, 75mm sides; Double convex lens; Double concave lens; Single convex lens; and Single concave lens.
  • Description:
    Large Magnetic Demonstration Prism Set