Tabletop Clamp and Support Rod
20" Rod, ⅜"
Catalog # 470019-516
Supplier: HUMBOLDT MFG CO
Número de proveedor #: H-21160
Jump to:
Product Details & Documents
Firm Tabletop Support.
- Sturdy cast-iron clamp
- Rod can be held vertically or horizontally
Especificaciones
- Length:20" Rod
- Rod Diameter:⅜"