Cladophora, General Slide
Cladophora, General, Whole mount, Fast Green and Mallory's
Catalog # 470180-968
Supplier: Avantor
Jump to:
Product Details & Documents
Filamentous Algae
- Identifying features clearly distinguishable
- Rigorous quality control standards
Branched filaments with multinucleate cells.
Specifications
- Description:Cladophora, General
- Stain:Fast Green and Mallory's
- Orientation:Whole mount