Orders LinkContactUsLinkComponent
Ceramic Fiber Squares
Ceramic Fiber Squares
Supplier:   ARMIL/CFS INC SE
Ceramic Fiber Squares
Supplier:  ARMIL/CFS INC SE
Certificates
Ceramic Fiber Squares
Ceramic Fiber Squares
1 Options Available Below
Product Details & Documents
Made of ceramic fiber with organic binder, for use under hot vessels to protect table tops. Does not contain asbestos. Package of 12.
Product Family Options
Product Information
  • Availability
    Price
  • 470122-536
    Ceramic Fiber Squares, 13 x 13 cm
    Ceramic Fiber Squares, 13 x 13 cm
    Log in for availability
    Log in for availability
    Paquete de 12
    Paquete de 12 - $1,124.64
    $1,124.64
    Paquete de 12
       $1,124.64
    Supplier Number: 550F-5X5
  • 470122-538
    Ceramic Fiber Squares, 15 x 15 cm
    Ceramic Fiber Squares, 15 x 15 cm
    Discontinued
    Supplier Number: 550F-6X6