Orders LinkContactUsLinkComponent
Cellophane
Cellophane
Cellophane
Cellophane
Use the various colors to demonstrate plants’ reaction to light waves.
  • Made from recycled materials
Also useful for demonstrating the effects of mixing various colors of light.
Product Family Options
ROL
  • Description
    Color
    Size
  • Cellophane Roll
    Red
    20×100"
    Each (1UN)
    Each (1UN) - $572.99
    $572.99
    Each (1UN)
       $572.99
     : 71002
  • Cellophane Roll
    Yellow
    20×100"
    Each (1UN)
    Each (1UN) - $572.99
    $572.99
    Each (1UN)
       $572.99
     : 71008
  • Cellophane Roll
    Blue
    20×100"
    Each (1UN)
    Each (1UN) - $572.99
    $572.99
    Each (1UN)
       $572.99
     : 71006
Sheets
  • Description
    Color
    Size
  • Cellophane Sheets
    Red
    20×60"
    Each (1UN)
    Each (1UN) - $125.40
    $125.40
    Each (1UN)
       $125.40