Cell Poster Series Set
Catalog # 470176-814
Proveedor: FILMS FOR THE HUMANITIES & SCI
Especificaciones
- Anchura:43 cm
- Descripción:Cell Poster Series Set
- Longitud:56 cm
- Referencia:470176-814
- Se ajuste:Laminated
- Nº de proveedor:10900TPOLA
Especificaciones
Acerca de este artículo
Comprehensive Coverage of All Types of Cells
- Spectacular 3-D illustrations highlight this unique set of posters
Each print depicts key areas of the cell biology curriculum in vivid color and stunning detail and is accompanied by descriptive text and labels. Posters include Cell Organization, Cell Shapes and Sizes, Mitosis, Meiosis, DNA, Plant Cells, Cell Membranes, Human Cells, Bacteria, and Organelles.