Orders LinkContactUsLinkComponent
Ward's® Cell Manipulative Models
Ward's® Cell Manipulative Models
Supplier:   Avantor
Ward's® Cell Manipulative Models
Supplier:  Avantor
Certificates
Ward's® Cell Manipulative Models
Ward's® Cell Manipulative Models
2 Options Available Below
Product Details & Documents
Students Create Their Own Cells
Introduce, reinforce, and test your students’ knowledge of cell structures and their functions with these fun and easy to use manipulative models. Using Velcro, students can attach organelles to the cell as they discover the fascinating complexity of plant and animal cells. Each model set comes with ten laminated cell sheets and ten sets of organelles, as well as a teacher’s guide with suggested activities.
Product Family Options
Información del producto
  • No. of Lab Groups
    Availability
    Price
  • 470003-984
    Cell Manipulative Models, Animal
    Cell Manipulative Models, Animal
    10
    Log in Para disponibilidad
    Log in Para disponibilidad
    Kit de 1
    Kit de 1 - $2,799.00
    $2,799.00
    Kit de 1
       $2,799.00
  • 470003-988
    Cell Manipulative Models, Plant
    Cell Manipulative Models, Plant
    10
    Log in Para disponibilidad
    Log in Para disponibilidad
    Kit de 1
    Kit de 1 - $2,072.40
    $2,072.40
    Kit de 1
       $2,072.40