Orders LinkContactUsLinkComponent
Cell Energy Posters
Cell Energy Posters
Cell Energy Poster Set, Laminated, 61 cm, 45.7 cm
Catalog # 470023-490
Supplier:   Avantor
Cell Energy Posters
Catalog # 470023-490
Supplier:  Avantor
Cell Energy Posters
Cell Energy Posters
Cell Energy Poster Set, Laminated, 61 cm, 45.7 cm
Price
$1,542.40
Log in Para disponibilidad
Kit de 1
Product Details & Documents
Illustrate How Plants and Animals Produce Energy
  • Laminated
Colorful, labeled diagrams and descriptive text highlight the most important structures and processes found in plant and animal cells in these two informative charts. The Photosynthesis chart features a detailed cross section of a chloroplast that is labeled with descriptive text discussing light reactions and the Calvin cycle. The Cell Respiration chart illustrates the gross structure of a mitochondria along with descriptive text that explains glycolysis, the Krebs cycle, and electron transport.
Especificaciones
  • Width:
    45.7 cm
  • Description:
    Cell Energy Poster Set
  • Length:
    61 cm
  • Finish:
    Laminated