Orders LinkContactUsLinkComponent
Ward's® Cell Concept Clings
Supplier:   Avantor
Ward's® Cell Concept Clings
Supplier:  Avantor
Certificates
Ward's® Cell Concept Clings
Ward's® Cell Concept Clings
2 Options Available Below
Product Details & Documents
Demonstrate and reinforce popular biological concepts with science that sticks.
Use these colorfully illustrated cling images to provide interactive learning while demonstrating and reinforcing popular biological concepts. The shapes are made from vinyl static cling material that will adhere to and easily remove from most smooth, shiny surfaces such as whiteboards and windows.
The Mitosis and Meiosis kit is aligned to NGSS standards (MS/HS: LS1, LS3) and is also available for purchase with a link to a NewPath digital lesson, which includes a variety of resources on the topic, including background information, illustrations and self-assessments.
Product Family Options
Información del producto
  • Availability
    Price
  • 470024-260
    Ward's® Cell Concept Clings, Mitosis and Meiosis
    Ward's® Cell Concept Clings, Mitosis and Meiosis
    Log in Para disponibilidad
    Log in Para disponibilidad
    Kit de 1
    Kit de 1 - $2,419.80
    $2,419.80
    Kit de 1
       $2,419.80
  • 470024-262
    Ward's® Cell Concept Clings, Plant and Animal Cell
    Ward's® Cell Concept Clings, Plant and Animal Cell
    Log in Para disponibilidad
    Log in Para disponibilidad
    Kit de 1
    Kit de 1 - $1,606.20
    $1,606.20
    Kit de 1
       $1,606.20
  • 470024-264
    Ward's® Cell Concept Clings, Genetics
    Ward's® Cell Concept Clings, Genetics
    Discontinued