Orders LinkContactUsLinkComponent
Cartridge-Type Deionizers
Cartridge-Type Deionizers
Certificates
Cartridge-Type Deionizers
Cartridge-Type Deionizers
2 Options Available Below
Product Details & Documents
Great for lab use.
  • Research model cartridge removes essentially all ionizable constituents including silica and free carbon dioxide
  • Universal model cartridge; does not remove silica or carbon dioxide
  • Single cartridge holder with faucet adapter (includes a brass control valve, inlet hose and outlet tubing), consists of one heavy-duty stamped steel mounting fixture, pressure fitting, tubing and washers
Transparent, plastic cartridges contain ion-exchange resins of the mixed-bed type for providing high purity water at low flow rates. For purification either by intermittent or steady flow. Intended for wall mounting, they are designed for operation at pressures up to 100 psi with a max flow of 27 liters per hour in the single cartridge holder.
Product Family Options
Product Information
  • Length
    Flow rate
    Availability
    Price
  • 470150-336
    Research cartridge, 1000 grains (as CaCO3) ion exchange capacity
    Research cartridge, 1000 grains (as CaCO3) ion exchange capacity
    20½"
    27 L/hr
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $6,723.19
    $6,723.19
    Log in for availability
    Case de 6 (1UN)
    Case de 6 (1UN) - $7,328.27
    $7,328.27
    Each (1UN)
       $6,723.19
    Case de 6 (1UN)
       $7,328.27
    Supplier Number: 1X-02-4030
  • 470150-334
    Universal cartridge, 1600 grains (as CaCO3) ion exchange capacity
    Universal cartridge, 1600 grains (as CaCO3) ion exchange capacity
    20½"
    27 L/hr
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $1,591.47
    $1,591.47
    Log in for availability
    Case de 6 (1UN)
    Case de 6 (1UN) - $1,734.70
    $1,734.70
    Each (1UN)
       $1,591.47
    Case de 6 (1UN)
       $1,734.70
    Supplier Number: 1X-02-4040
  • Discontinued
    Supplier Number: -