Orders LinkContactUsLinkComponent
Cartesian Diver
Cartesian Diver
Cartesian Diver, 4 cm
  470235-702
Supplier #:  305-10
Cartesian Diver
  470235-702
Cartesian Diver
Cartesian Diver
Cartesian Diver, 4 cm
$114.89
Each (1UN) Paquete de 10 (1UN)
Ideal for Showing Archimedes’ Principle
Students will be fascinated at their first encounter with our Cartesian Diver. We supply a hollow glass diver, even shaped like a diver, with a 10" x 1 1/2" glass cylinder, a rubber sheet and rubber band. Fill the cylinder with water, drop in the diver so he floats, fasten the sheet tautly across the top and then gently depress the sheet. The diver will dive, his speed depending upon the pressure you place on the sheet; release the sheet and the diver returns to the surface. The diver’s behavior illustrates the uniform transmission of pressure in all directions, the compressibility of gases and Archimedes’ Principle.
  • Description:
    Cartesian Diver
  • Length:
    4 cm