Cardiac Muscle, Thin Section Slide
Cardiac Muscle, Thin Section Slide, Thin section, Iron Hematoxylin
470183-666
: Avantor
Extremely Thin Plastic Embedded Tissue For Greater Viewing Clarity
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Shows branching fibers, striations, and nuclei.
- Description:Cardiac Muscle, Thin Section Slide
- Stain:Iron Hematoxylin
- Orientation:Thin section