Camelid Blood Slide
Camelid blood microscope slide, Smear, Giemsa
Catalog # 470182-766
Supplier: Avantor
Jump to:
Product Details & Documents
There is no nucleus in the oval-shaped erythrocytes, which can swell up to 240% of their normal size without bursting. From llama.
Especificaciones
- Description:Camelid blood microscope slide
- Stain:Giemsa
- Orientation:Smear