Orders LinkContactUsLinkComponent
Ward's® Calcite (Iceland Spar)
Ward's® Calcite (Iceland Spar)
Supplier:   Avantor
Ward's® Calcite (Iceland Spar)
Supplier:  Avantor
Certificates
Ward's® Calcite (Iceland Spar)
Ward's® Calcite (Iceland Spar)
2 Options Available Below
Product Details & Documents
Clear rhombs showing double refraction.
  • Collected in Mexico
  • Multiple sizes and pack options to fit specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" - 2×3". Packaged in a labeled storage box for easy organization.
All Specimen dimensions are considered an average size range.
Product Family Options
Specimens
Información del producto
  • Specimen Dimensions
    Availability
    Price
  • 470025-518
    Calcite (Iceland Spar), student specimens
    Calcite (Iceland Spar), student specimens
    1×1 - 1×1"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $832.78
    $832.78
    Paquete de 10
       $832.78
  • 470025-522
    Calcite (Iceland Spar), bulk pack, 1 kg
    Calcite (Iceland Spar), bulk pack, 1 kg
    Size will vary
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,006.30
    $1,006.30
    Each (1UN)
       $1,006.30