Orders LinkContactUsLinkComponent
Ward's® Calcite (Cleavage)
Ward's® Calcite (Cleavage)
Supplier:   Avantor
Ward's® Calcite (Cleavage)
Supplier:  Avantor
Certificates
Ward's® Calcite (Cleavage)
Ward's® Calcite (Cleavage)
4 Options Available Below
Product Details & Documents
Pure translucent to white, rhombic cleavages, Mohs’ hardness of 3.
  • Collected in Mexico
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" - 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Información del producto
  • Specimen Dimensions
    Availability
    Price
  • 470025-512
    Calcite (Cleavege), bulk pack, 1 kg
    Calcite (Cleavege), bulk pack, 1 kg
    Size will vary
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $548.10
    $548.10
    Each (1UN)
       $548.10
  • 470025-514
    Calcite (Cleavage), hand specimen, each
    Calcite (Cleavage), hand specimen, each
    2×3 - 3×4"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $353.95
    $353.95
    Each (1UN)
       $353.95
  • 470025-510
    Calcite (Cleavage), student specimen
    Calcite (Cleavage), student specimen
    1×1 - 1×1"
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 10
    Paquete de 10 - $344.80
    $344.80
    Paquete de 10
       $344.80
  • 470024-924
    Calcite thin section
    Calcite thin section
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,045.20
    $1,045.20
    Each (1UN)
       $1,045.20