Orders LinkContactUsLinkComponent
C Clamps
C Clamps
C Clamps
Certificates
C Clamps
2 Options Available Below
Product Details & Documents
Securely hold two flat object together.
  • Cast iron frame
  • Screw thread mechanism
These cast iron C clamps help to securely holes surfaces together. The maximum opening of the jaw needs to be taken into account when selecting the correct size. The horizontal torque bar allows you to apply a lot of force to tighten or release.
Product Family Options
Información del producto
  • Availability
    Price
  • 470005-368
    8 cm (3") Jaw Opening
    8 cm (3") Jaw Opening
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $165.77
    $165.77
    Each (1UN)
       $165.77
    Supplier Number: CLMPC3
  • 470104-094
    10 cm (4") Jaw Opening
    10 cm (4") Jaw Opening
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $215.86
    $215.86
    Each (1UN)
       $215.86
    Supplier Number: CLMPC4