Ward's® Butterfly Life Cycle Poster
470023-244
: Avantor
- Anchura:86 cm
- Descripción:Butterfly Life Cycle Poster
- Longitud:53 cm
- Referencia:470023-244
- Se ajuste:Unlaminated
- Nº de proveedor:WARD470023-244
Clearly Represents All Stages of Metamorphosis
- A variety of other North American butterfly species are also represented
The life cycle of the colorful, widely recognized Monarch butterfly is represented in this large, striking poster. The four stages of metamorphosis: egg, larva, pupa, and adult, are depicted in nine vivid, close-up photographs.