Orders LinkContactUsLinkComponent
Burner Wing Top
Burner Wing Top
Wing top for burners with flame stablizer
Catalog # 470148-920
Supplier:   VWR International
Burner Wing Top
Catalog # 470148-920
Supplier:  VWR International
Burner Wing Top
Burner Wing Top
Wing top for burners with flame stablizer
Price
$153.36
Log in for availability
Each (1UN) Case de 10 (1UN)
Product Details & Documents
It provides a broad, flat flame and fits mixing tubes with 1/2" outer diameter without stabilizer or 11/16" with stabilizer.
Specifications
  • Description:
    Wing top for burners with flame stablizer