Orders LinkContactUsLinkComponent
Burner Tripods
Burner Tripods
Supplier:   VWR International
Burner Tripods
Supplier:  VWR International
Certificates
Burner Tripods
2 Options Available Below
Product Details & Documents
Good quality, enameled cast iron tripods for supporting small objects over a burner.
  • Cast iron, enameled finish
  • Removable zinc plated legs
Good quality, enameled cast iron tripods for supporting small objects over a burner. The legs detach for easy storage.
Product Family Options
Product Information
  • Ext.Ø
    Availability
    Price
  • 470148-880
    Tripod, Burner, 23 cm x 13 cm x 9 cm
    Tripod, Burner, 23 cm x 13 cm x 9 cm
    5" (3.5" I.D.)
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $7.20
    $7.20
    Clearance
    Log in for availability
    Case de 20 (1UN)
    Case de 20 (1UN) - $144.00
    $144.00
    Clearance
    Each (1UN)
       $7.20
    Case de 20 (1UN)
       $144.00
  • 470104-578
    Tripod, Burner, 23 cm x 11 cm x 9 cm
    Tripod, Burner, 23 cm x 11 cm x 9 cm
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $298.49
    $298.49
    Each (1UN)
       $298.49