Buret Brushes
Buret Brushes, 3", 3/4", 36"
Catalog # 470177-964
Supplier: ALLIED K & R
Número de proveedor #: D210
Jump to:
Product Details & Documents
The long handle and fan tip is ideal for cleaning 50 mL burets. Size: 36"L; brush size: 3/4" dia. x 3"L. Package of 12.
Especificaciones
- Brush head Ø:3/4"
- Description:Buret Brushes
- Total length:36"
- Brush Head Length:3"