Bullet ID Poster
Bullet ID Poster
Catalog # 470023-270
Supplier: PRECISION FORENSICS TESTING
Número de proveedor #: PB-01
Jump to:
Product Details & Documents
Classify Firearms From Bullet Appearance
- Size: 24" x 36"
Focusing on bullet comparison and the marks associated with the comparisons, this poster outlines how to identify firearms involved in crimes from looking at the bullets fired. It also demonstrates some of the different class characteristics of bullets. Size: 24" x 36".
Especificaciones
- Description:Bullet ID Poster