Brown Adipose Tissue Slide
Brown Adipose Tissue, Section, Hematoxylin and Eosin
Catalog # 470182-630
Supplier: Avantor
Jump to:
Product Details & Documents
Thermoregulation is Primary Function
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. From the so-called “hibernating gland” of rodents. In these cells the lipid droplets do not become confluent.
Especificaciones
- Description:Brown Adipose Tissue
- Stain:Hematoxylin and Eosin
- Orientation:Section