Orders LinkContactUsLinkComponent
Bridge Building Contest Kit
Bridge Building Contest Kit
Supplier:   Avantor
Bridge Building Contest Kit
Supplier:  Avantor
Certificates
Bridge Building Contest Kit
Bridge Building Contest Kit
2 Options Available Below
Product Details & Documents
Cutting Edge Construction and Physical Engineering
  • Physics in a contest setting
  • Create a variety of bridges
  • Learn about force and stress
Applying physics concepts to construction in a contest setting, students create a variety of different bridges — including beam, truss, and arch styles. With each build, students learn about the forces and stresses their creations must endure and develop a greater understanding of structural integrity and the physics of bridge construction. The added motivation of a national competition encourages students to thoroughly research and design the most structurally sound bridges.
These contest kits, developed in partnership with Brookhaven National Laboratories, and aligned with national standards, are available in two versions to meet the specific needs of your classroom and students. The basswood version is designed primarily for high school students. The straw building kit is ideal for introducing middle school students to the basics of design and structure without the safety concerns of sharp instruments or glue. Both sets include materials for 5 student groups.
Product Family Options
Información del producto
  • Availability
    Price
  • 470003-056
    Bridge Building Contest Kit, Basswood
    Bridge Building Contest Kit, Basswood
    Log in Para disponibilidad
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $6,638.67
    $6,638.67
    Log in Para disponibilidad
    Paleta de 15 (1UN)
    Paleta de 15 (1UN) - $99,579.98
    $99,579.98
    Each (1UN)
       $6,638.67
    Paleta de 15 (1UN)
       $99,579.98
  • 470003-058
    Bridge Building Contest Kit, Straws
    Bridge Building Contest Kit, Straws
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,126.80
    $1,126.80
    Each (1UN)
       $1,126.80