Orders LinkContactUsLinkComponent
Box Magnifiers
Box Magnifiers
Box Magnifiers
Certificates
Box Magnifiers
2 Options Available Below
Product Details & Documents
Contains and Magnifies Small Specimens
These clear plastic boxes feature a magnifying press-fit lid; simply place the specimen to be studied in the box, cover, and observe. They can also be used when permanently embedding small specimens in resin. Size: 4X magnifier 1 L×1 W×7/8H"; 3X magnifier 1 1/2 L×1 1/2 W×1 1/2 H".
Product Family Options
Product Information
  • Magnification
    Availability
    Price
  • 470016-876
    Box Magnifier
    Box Magnifier
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $18.14
    $18.14
    Log in for availability
    Caja de 250 (1UN)
    Caja de 250 (1UN) - $19.77
    $19.77
    Each (1UN)
       $18.14
    Caja de 250 (1UN)
       $19.77
    Supplier Number: 6455000
  • 470016-878
    Box Magnifier
    Box Magnifier
    Log in for availability
    Log in for availability
    Log in for availability
    Each (1UN)
    Each (1UN) - $21.84
    $21.84
    Log in for availability
    Caja de 250 (1UN)
    Caja de 250 (1UN) - $23.81
    $23.81
    Each (1UN)
       $21.84
    Caja de 250 (1UN)
       $23.81
    Supplier Number: 6455100