Graduated Cylinders with Plastic Base
Supplier: UNITED SCIENTIFIC SUPPLIES
Acerca de este artículo
Durable Borosilicate Glass Construction.
- Graduations permanently fused into glass
- Hexagonal base and bumper guard add protection for all sizes
Double metric scale, calibrated to contain. Borosilicate glass cylinders with pour spouts feature heavy uniform wall tubing.
5 Options Available Below
Product Details & Documents
Durable Borosilicate Glass Construction.
- Graduations permanently fused into glass
- Hexagonal base and bumper guard add protection for all sizes
Double metric scale, calibrated to contain. Borosilicate glass cylinders with pour spouts feature heavy uniform wall tubing.
Specifications for Product Family
Especificaciones
- Catalog No:
- 470188-950
- 470188-952
- 470188-954
- 470188-955
- 470121-934
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
Especificaciones
Product Family Options
Información del producto
- CapacityGraduation IntervalAvailabilityPrice
- 10 ml0.1 mlEach (1UN)$126.85Paquete de 10 (1UN)$138.27Supplier Number: CY137-10Especificaciones:
More
Specifications
Cat. No.470188-950FormTall - 25 ml0.5 mlEach (1UN)$162.30Paquete de 10 (1UN)$176.91Supplier Number: CY137-25Especificaciones:
More
Specifications
Cat. No.470188-952FormTall - 50 ml1 mlEach (1UN)$228.97Paquete de 10 (1UN)$249.57Supplier Number: CY137-50Especificaciones:
More
Specifications
Cat. No.470188-954FormTall - 50 ml1 mlEach (1UN)$228.97Paquete de 10 (1UN)$249.57Supplier Number: CY137-50Especificaciones:
More
Specifications
Cat. No.470188-955FormTall - Supplier Number: CY137-100Especificaciones:
More
Specifications
Cat. No.470121-934FormTall