Orders LinkContactUsLinkComponent
Borosilicate Capillary Melting Point Tubes
Borosilicate Capillary Melting Point Tubes
Borosilicate Capillary Melting Point Tubes
Certificates
Borosilicate Capillary Melting Point Tubes
Borosilicate Capillary Melting Point Tubes
4 Options Available Below
Product Details & Documents
Intermediate hardness and melting point.
Packaging: 100 tubes per vial.
Product Family Options
Información del producto
  • Length
    Wall thickness
    Availability
    Price
  • 470122-726
    Capillary tubes, open both ends
    Capillary tubes, open both ends
    100 mm (4")
    0.2 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $536.76
    $536.76
    Each (1UN)
       $536.76
    Supplier Number: VWR-WLS44845
  • 470122-730
    Capillary tubes, open one end
    Capillary tubes, open one end
    100 mm (4")
    0.2 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $623.63
    $623.63
    Each (1UN)
       $623.63
    Supplier Number: VWR-WLS44846
  • 470122-728
    Capillary tubes, open both ends
    Capillary tubes, open both ends
    150 mm (6")
    0.2 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $539.94
    $539.94
    Paquete de 100
       $539.94
    Supplier Number: FD-15185-OE
  • 470122-732
    Capillary tubes, open one end
    Capillary tubes, open one end
    150 mm (6")
    0.2 mm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $554.85
    $554.85
    Paquete de 100
       $554.85
    Supplier Number: FD15185-CE