Bone, Ground Preparation Slide
Bone, Ground Preparation, Longitudinal section, Unstained
Catalog # 470015-864
Supplier: Avantor
Jump to:
Product Details & Documents
Examine bone tissue microscopically
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Thin section showing lacunae distinct with canaliculi.
Specifications
- Description:Bone, Ground Preparation
- Stain:Unstained
- Orientation:Longitudinal section