Ward's® Compact Bone Slide
Bone, Compact Slide, Cross section & longitudinal section, Hematoxylin and Eosin
Catalog # 470182-656
Supplier: Avantor
Jump to:
Product Details & Documents
Rigid, dense layer of bone
- Identifying features clearly distinguishable
- Rigorous quality control standards
Mammal. Decalcified and stained.
Specifications
- Description:Bone, Compact Slide
- Stain:Hematoxylin and Eosin
- Orientation:Cross section & longitudinal section