ContactUsLinkComponent
PYREX® Round Bottom Boiling Flasks
  470202-168
Product available on GSA Advantage®
undefined
PYREX® Round Bottom Boiling Flasks
  470202-168
 :  4280-1L
Product available on GSA Advantage®
  • Descripción:
    Flask, boiling, Pyrex®, 1000 ml, w/ round bottom, Corning
  • Referencia:
    470202-168
  • Nº de proveedor:
    4280-1L
Recommended for critical boiling applications.
These round bottom flasks are made from quality borosilicate and have a heavy neck rim to minimize breakage.