BIO-FOAM® Impression Foam
BIO-FOAM® Impression Foam, 6", 2¼", 14"
Catalog # 470020-454
Supplier: FORENSICS SOURCE
Número de proveedor #: 1005733
Jump to:
Product Details & Documents
A Clean, Quick, Easy Way to Make Footprint Impressions
Follow in the footsteps of actual forensic detectives with this professional field technique. Press any footwear into the blue BIO-FOAM®, and after removal, the foam retains a detailed footprint impression, which can be stored in the original cardboard container. For further study, a cast can be made of foam impressions by coating with hair spray and pouring in a prepared casting material (available separately). Designed for one use per BIO-FOAM®. Size: 14"L x 6"W x 2 1/4"H
Especificaciones
- Description:BIO-FOAM® Impression Foam
- Height:2¼"
- Length:14"
- Width:6"