Bibulous Paper
Ward's® Bibulous Paper, 4×6"
Catalog # 470005-600
Supplier: RAYLABCON INC SE
Número de proveedor #: 20 10 5000
Jump to:
Product Details & Documents
Common blotting and cleaning paper.
- Quality slide making supplies
- Helpful for maintaining cleanliness of slides
Use this bibulous paper to clean glass surfaces of dust and lint without leaving streaks. It also blots excess moisture from slides containing living materials. Staple-bound booklet of 50 sheets.
Especificaciones
- Description:Ward's® Bibulous Paper
- Dimensions:4×6"