Berge's Manual Of Determinative Bacteriology: Ninth Edition
Catalog # 470002-378
Supplier: LIPPINCOTT WILLIAMS & WILKINS
Specifications
- Descripción:Berge's Manual Of Determinative Bacteriology: 9Th Edition
- Longitud:22 cm
- Referencia:470002-378
- No. of Pages:787
- Anchura:28 cm
- Nº de proveedor:0683006037
Specifications
About this item
John G. Holt et al
A reference to aid in the identification of unknown bacteria. It represents all of the bacteria that have been cultured and provides taxonomic keys to distinguish between unknown cultures.