Orders LinkContactUsLinkComponent
A-Frame Base with Rod
A-Frame Base with Rod
A-Frame Base with Rod
Certificates
A-Frame Base with Rod
A-Frame Base with Rod
1 Options Available Below
Product Details & Documents
These cast-iron bases have three-point stability on nearly any surface.
  • Rod and base set consisting of an A-shaped retort base
  • Baked enamel finish
  • Stackable even with rod attached
The retort stand is made of corrosion resistant, powder coated pressed steel, and features a tapped hole, suitable for connecting the included rod, or any other rods with a 10×1.5 mm metric thread. Heavy weight construction provides superb stability for basic experiments and setups.
Product Family Options
Información del producto
  • Base Dimensions
    Rod Diameter
    Rod Length
    Availability
    Price
  • 180×180 mm
    9.5 mm
    44 cm
    Discontinued
    Supplier Number: ABSE12
  • 220×220 mm
    12.7 mm
    59 cm
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $1,425.28
    $1,425.28
    Each (1UN)
       $1,425.28
    Supplier Number: ABSE12SPL