Barnes Dropping Bottle
30 mL
Catalog # 470123-892
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: BOT030-PK/36
Jump to:
Product Details & Documents
Barnes Dropping Bottles are made of heavy, clear glass.
- Heavy, Clear Glass Construction
- Glass Pipet and Rubber Bulb/Stopper Included
- Square
Each bottle includes a glass pipet.
A rubber bulb with a stopper is also included.
Especificaciones
- Description:30 mL