Bare Nichrome Wire
The Bare Nichrome Wire can handle temperatures of up to 1010°C (1850°F).
- Alloy of Nickel and Chromium
- Grams: 113
- Wire Can Handle Temperatures up to 1010°C (1850°F)
- High Performance
- Ideal for Rheostats and Controls Requiring Compact Resistance
It's particularly suited to heavy duty rheostats and controls that require compact resistance.
The wire's alloy is made of nickel and chromium.
The Bare Nichrome Wire can handle temperatures of up to 1010°C (1850°F).
- Alloy of Nickel and Chromium
- Grams: 113
- Wire Can Handle Temperatures up to 1010°C (1850°F)
- High Performance
- Ideal for Rheostats and Controls Requiring Compact Resistance
It's particularly suited to heavy duty rheostats and controls that require compact resistance.
The wire's alloy is made of nickel and chromium.
Product Family Options
- DescriptionLength
- : D24
- : D26
- : D20
- : D-28
- : 4 OZ
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.