Orders LinkContactUsLinkComponent
Bare Copper Wire, 113 g Spool
Bare Copper Wire, 113 g Spool
Bare Copper Wire, 113 g Spool
Certificates
Bare Copper Wire, 113 g Spool
Bare Copper Wire, 113 g Spool
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A bare copper wire of the highest quality. Available in several standard AWG sizes.
  • 113 g spool
Product Family Options
Información del producto
  • Availability
    Price
  • 470005-942
    Wire, Bare Copper, 30 Gauge, 259 m, 113 grams
    Wire, Bare Copper, 30 Gauge, 259 m, 113 grams
    Log in Para disponibilidad
    Log in Para disponibilidad
    Each (1UN)
    Each (1UN) - $6.20
    $6.20
    Liquidación
    Each (1UN)
       $6.20
    Supplier Number: B30
  • 470201-430
    Bare Copper Wire — 113g Spool
    Bare Copper Wire — 113g Spool
    Discontinued
    Supplier Number: B20
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 