Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Descripción:Bacterial Pollution of Water Lab Activity
- No. of Lab Groups:15
- Referencia:470023-912
- Grado:HS
- Nº de proveedor:1400
Specifications
About this item
Use the Exact Water-Testing Procedures Used by Health Authorities
- Collect your own samples
- Practice preparing microscope slides
- Includes a teacher's guide
Students will collect water samples, extract a few drops, and add them to an agar medium. They are able to see and study the bacterial colonies that form as a result. The kit contains all materials needed for 15 setups, including a teacher’s manual and student instructions.
The kit contains all materials needed for 15 setups, including a teacher’s manual and student instructions. Designed for environmental science, microbiology, and biology courses.