Bacillus megaterium Slide
Bacillus megaterium, Smear, Gram (+)
470180-862
: Avantor
Bacteria often found in soil
- Identifying features clearly distinguishable
- Rigorous quality control standards
Rods. Endospores formed.
- Description:Bacillus megaterium
- Stain:Gram (+)
- Orientation:Smear