Orders LinkContactUsLinkComponent
Ward's® Azurite
 :   Avantor
Ward's® Azurite
 :  Avantor
Ward's® Azurite
Ward's® Azurite
Bright blue crystalline veins and coatings in a matrix of mixed copper ore.
  • Multiple sizes and pack options to fit specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" - 2×3". Packaged in a labeled storage box for easy organization.
All Specimen dimensions are considered an average size range.
Product Family Options
Specimens
  • Description
    Specimen Dimensions
  • Azurite, student specimens
    1×1 - 1×1"
    Paquete de 10
    Paquete de 10 - $483.40
    $483.40
    Paquete de 10
       $483.40
  • Azurite, bulk pack, 1 kg
    Size will vary
    Each (1UN)
    Each (1UN) - $872.28
    $872.28
    Each (1UN)
       $872.28
  • Azurite, hand specimen, Each
    2×3" - 3×4"
    Each (1UN)
    Each (1UN) - $585.33
    $585.33
    Each (1UN)
       $585.33